Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTGTTGGTCCTTCGGCTGCCATG[A/G]ACCCAGGATAAGACATCTACTTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612342 | ||||||||||||||||||||
Literature Links: |
GGT7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GGT7 - gamma-glutamyltransferase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178026.2 | 1427 | Silent Mutation | GTC,GTT | V,V 635 | NP_821158.2 | |
XM_011528782.2 | 1427 | Silent Mutation | GTC,GTT | V,V 401 | XP_011527084.1 | |
XM_011528783.2 | 1427 | Silent Mutation | GTC,GTT | V,V 352 | XP_011527085.1 | |
XM_017027809.1 | 1427 | UTR 3 | XP_016883298.1 | |||
XM_017027810.1 | 1427 | Intron | XP_016883299.1 | |||
XM_017027811.1 | 1427 | Intron | XP_016883300.1 |