Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATTTCATCTCCTTCCCTGCAGAA[C/T]CCCAAGGAGTGGACCAAGAAGGTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613599 MIM: 138550 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABHD12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABHD12 - abhydrolase domain containing 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042472.2 | 2491 | Intron | NP_001035937.1 | |||
NM_015600.4 | 2491 | Intron | NP_056415.1 | |||
XM_005260698.1 | 2491 | Intron | XP_005260755.1 | |||
XM_011529214.2 | 2491 | Intron | XP_011527516.1 | |||
XM_011529215.1 | 2491 | Intron | XP_011527517.1 | |||
XM_011529218.1 | 2491 | Intron | XP_011527520.1 | |||
XM_017027796.1 | 2491 | Intron | XP_016883285.1 | |||
XM_017027797.1 | 2491 | Intron | XP_016883286.1 | |||
XM_017027798.1 | 2491 | Intron | XP_016883287.1 |
PYGB - phosphorylase, glycogen; brain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002862.3 | 2491 | Silent Mutation | AAC,AAT | N,N 794 | NP_002853.2 |