Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGACTGTCCTGCCACCGCACCAGG[A/G]GGAAGTCTTGAAATGTGAGTCTGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608270 | ||||||||||||||||||||
Literature Links: |
TASP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TASP1 - taspase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323602.1 | 915 | Silent Mutation | CCC,CCT | P,P 266 | NP_001310531.1 | |
NM_001323603.1 | 915 | Silent Mutation | CCC,CCT | P,P 295 | NP_001310532.1 | |
NM_001323604.1 | 915 | Silent Mutation | CCC,CCT | P,P 295 | NP_001310533.1 | |
NM_017714.2 | 915 | Silent Mutation | CCC,CCT | P,P 397 | NP_060184.2 | |
XM_011529292.1 | 915 | Silent Mutation | CCC,CCT | P,P 295 | XP_011527594.1 | |
XM_011529294.1 | 915 | Silent Mutation | CCC,CCT | P,P 295 | XP_011527596.1 | |
XM_017027927.1 | 915 | Silent Mutation | CCC,CCT | P,P 295 | XP_016883416.1 | |
XM_017027928.1 | 915 | Silent Mutation | CCC,CCT | P,P 295 | XP_016883417.1 | |
XM_017027929.1 | 915 | Intron | XP_016883418.1 | |||
XM_017027930.1 | 915 | Silent Mutation | CCC,CCT | P,P 211 | XP_016883419.1 | |
XM_017027931.1 | 915 | Intron | XP_016883420.1 |