Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGTGGGTGGCAATGCTGTGACA[C/T]TGACCGAGTACATATCAGCCCAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606411 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OCSTAMP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OCSTAMP - osteoclast stimulatory transmembrane protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC13A3 - solute carrier family 13 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011554.2 | 1624 | Missense Mutation | AAT,AGT | N,S 539 | NP_001011554.1 | |
NM_001193339.1 | 1624 | Missense Mutation | AAT,AGT | N,S 536 | NP_001180268.1 | |
NM_001193340.1 | 1624 | Missense Mutation | AAT,AGT | N,S 504 | NP_001180269.1 | |
NM_001193342.1 | 1624 | Missense Mutation | AAT,AGT | N,S 488 | NP_001180271.1 | |
NM_022829.5 | 1624 | Missense Mutation | AAT,AGT | N,S 586 | NP_073740.2 |