Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGACAGGGTTGACAGCCGAGTTG[A/G]CCCACAGGAGCCAGAAGGAGGTTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604525 | ||||||||||||||||||||
Literature Links: |
HRH3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HRH3 - histamine receptor H3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007232.2 | 1208 | Missense Mutation | GCC,GTC | A,V 403 | NP_009163.2 | |
XM_005260266.3 | 1208 | Missense Mutation | GCC,GTC | A,V 403 | XP_005260323.1 | |
XM_017027623.1 | 1208 | Missense Mutation | GCC,GTC | A,V 349 | XP_016883112.1 |