Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602548 MIM: 605071 MIM: 604784 | ||||||||||||||||||||
Literature Links: |
LKAAEAR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LKAAEAR1 - LKAAEAR motif containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6813 - microRNA 6813 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OPRL1 - opioid related nociceptin receptor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RGS19 - regulator of G-protein signaling 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039467.1 | 434 | Missense Mutation | CAG,CGG | Q,R 56 | NP_001034556.1 | |
NM_005873.2 | 434 | Missense Mutation | CAG,CGG | Q,R 56 | NP_005864.1 | |
XM_005260182.3 | 434 | Missense Mutation | CAG,CGG | Q,R 34 | XP_005260239.1 | |
XM_005260183.2 | 434 | Missense Mutation | CAG,CGG | Q,R 34 | XP_005260240.1 | |
XM_011528484.1 | 434 | Missense Mutation | CAG,CGG | Q,R 85 | XP_011526786.1 | |
XM_011528485.2 | 434 | Missense Mutation | CAG,CGG | Q,R 56 | XP_011526787.1 | |
XM_011528486.2 | 434 | Missense Mutation | CAG,CGG | Q,R 56 | XP_011526788.1 |
TCEA2 - transcription elongation factor A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |