Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTGGGCCCTCGGGGCTCAGTCAT[C/T]GGCCAGGGTGATGACGTCGTACTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610827 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCIF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PCIF1 - PDX1 C-terminal inhibiting factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022104.3 | 3915 | Intron | NP_071387.1 | |||
XM_011528980.2 | 3915 | Intron | XP_011527282.1 | |||
XM_011528981.2 | 3915 | Intron | XP_011527283.1 | |||
XM_017028013.1 | 3915 | Intron | XP_016883502.1 | |||
XM_017028014.1 | 3915 | Intron | XP_016883503.1 |
ZNF335 - zinc finger protein 335 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022095.3 | 3915 | Missense Mutation | AAT,GAT | N,D 1341 | NP_071378.1 | |
XM_005260504.4 | 3915 | Missense Mutation | AAT,GAT | N,D 1340 | XP_005260561.1 | |
XM_011528979.2 | 3915 | Intron | XP_011527281.1 | |||
XM_017028012.1 | 3915 | Missense Mutation | AAT,GAT | N,D 1165 | XP_016883501.1 |