Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGCCTTGCTGACTCCCACCTTCC[A/G]GCCGGTGGAGCCCCGGGCCCCCTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610866 | ||||||||||||||||||||
Literature Links: |
UCKL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UCKL1-AS1 - UCKL1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF512B - zinc finger protein 512B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020713.2 | 2775 | Missense Mutation | CGG,TGG | R,W 882 | NP_065764.1 | |
XM_005260226.3 | 2775 | Missense Mutation | CGG,TGG | R,W 896 | XP_005260283.1 | |
XM_011528929.2 | 2775 | Missense Mutation | CGG,TGG | R,W 882 | XP_011527231.1 | |
XM_011528930.2 | 2775 | Missense Mutation | CGG,TGG | R,W 863 | XP_011527232.1 | |
XM_011528931.2 | 2775 | Missense Mutation | CGG,TGG | R,W 552 | XP_011527233.1 | |
XM_011528932.2 | 2775 | Missense Mutation | CGG,TGG | R,W 551 | XP_011527234.1 | |
XM_011528933.2 | 2775 | Intron | XP_011527235.1 |