Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTCCAGGAGGTCAGCCCGGTCCC[A/G]GCCCTGGAAGGCACAGTGCAGACAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZFP64 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZFP64 - ZFP64 zinc finger protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319146.1 | 1420 | Missense Mutation | CGG,TGG | R,W 258 | NP_001306075.1 | |
NM_018197.2 | 1420 | Intron | NP_060667.2 | |||
NM_022088.4 | 1420 | Intron | NP_071371.3 | |||
NM_199426.1 | 1420 | Intron | NP_955458.1 | |||
NM_199427.2 | 1420 | Missense Mutation | CGG,TGG | R,W 477 | NP_955459.2 | |
XM_005260449.1 | 1420 | Intron | XP_005260506.1 | |||
XM_006723822.1 | 1420 | Intron | XP_006723885.1 | |||
XM_011528900.1 | 1420 | Intron | XP_011527202.1 | |||
XM_017027945.1 | 1420 | Missense Mutation | CGG,TGG | R,W 475 | XP_016883434.1 | |
XM_017027946.1 | 1420 | Intron | XP_016883435.1 | |||
XM_017027947.1 | 1420 | Missense Mutation | CGG,TGG | R,W 423 | XP_016883436.1 | |
XM_017027948.1 | 1420 | Intron | XP_016883437.1 | |||
XM_017027949.1 | 1420 | Intron | XP_016883438.1 |