Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTCTCTGCTCACCTTGCCTTATC[A/G]TGTTGTTGATATCTTTCAAACTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116957 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RBL1 - RB transcriptional corepressor like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323281.1 | 2957 | Missense Mutation | ACG,ATG | T,M 593 | NP_001310210.1 | |
NM_001323282.1 | 2957 | Missense Mutation | ACG,ATG | T,M 593 | NP_001310211.1 | |
NM_002895.4 | 2957 | Missense Mutation | ACG,ATG | T,M 1020 | NP_002886.2 | |
NM_183404.3 | 2957 | Intron | NP_899662.1 | |||
XM_006723842.2 | 2957 | Missense Mutation | ACG,ATG | T,M 848 | XP_006723905.1 | |
XM_011528955.2 | 2957 | Missense Mutation | ACG,ATG | T,M 965 | XP_011527257.1 | |
XM_011528958.2 | 2957 | Intron | XP_011527260.1 | |||
XM_017027992.1 | 2957 | Missense Mutation | ACG,ATG | T,M 916 | XP_016883481.1 |