Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAGTACCAGGCTTTCCGGGAGGC[C/T]ATCACTGAGGCGGTGGAGGCACCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604699 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARFRP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARFRP1 - ADP ribosylation factor related protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134758.3 | 425 | Intron | NP_001128230.1 | |||
NM_001267544.2 | 425 | Intron | NP_001254473.1 | |||
NM_001267545.2 | 425 | Intron | NP_001254474.1 | |||
NM_001267546.2 | 425 | Intron | NP_001254475.1 | |||
NM_001267547.2 | 425 | Intron | NP_001254476.1 | |||
NM_001267548.2 | 425 | Intron | NP_001254477.1 | |||
NM_001267549.2 | 425 | Intron | NP_001254478.1 | |||
NM_003224.5 | 425 | Intron | NP_003215.1 | |||
XM_011528482.2 | 425 | Intron | XP_011526784.1 | |||
XM_011528483.1 | 425 | Intron | XP_011526785.1 | |||
XM_017027576.1 | 425 | Intron | XP_016883065.1 |
ZGPAT - zinc finger CCCH-type and G-patch domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083113.1 | 425 | Silent Mutation | GCC,GCT | A,A 85 | NP_001076582.1 | |
NM_001195653.1 | 425 | Silent Mutation | GCC,GCT | A,A 85 | NP_001182582.1 | |
NM_001195654.1 | 425 | Silent Mutation | GCC,GCT | A,A 85 | NP_001182583.1 | |
NM_032527.4 | 425 | Silent Mutation | GCC,GCT | A,A 85 | NP_115916.3 | |
NM_181485.2 | 425 | Silent Mutation | GCC,GCT | A,A 85 | NP_852150.2 |