Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCGACAGGCCAGAGTGTCAATGCG[C/T]CCCCTGCTATCCAGCCATTGGGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF341 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF341 - zinc finger protein 341 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282933.1 | 383 | Missense Mutation | CCC,TCC | P,S 41 | NP_001269862.1 | |
NM_001282935.1 | 383 | Silent Mutation | CGC,CGT | R,R 16 | NP_001269864.1 | |
NM_032819.4 | 383 | Missense Mutation | CCC,TCC | P,S 41 | NP_116208.3 |