Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCTGTTTGCAGAGGAGCTGGTGC[A/T]TGCAGGCTGGGAGAAGTGCTGGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610827 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107985388 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC107985388 - pre-mRNA-splicing factor cwc22-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCIF1 - PDX1 C-terminal inhibiting factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022104.3 | 415 | Missense Mutation | CAT,CTT | H,L 46 | NP_071387.1 | |
XM_011528980.2 | 415 | Missense Mutation | CAT,CTT | H,L 46 | XP_011527282.1 | |
XM_011528981.2 | 415 | Missense Mutation | CAT,CTT | H,L 46 | XP_011527283.1 | |
XM_017028013.1 | 415 | Missense Mutation | CAT,CTT | H,L 46 | XP_016883502.1 | |
XM_017028014.1 | 415 | UTR 5 | XP_016883503.1 |
ZNF335 - zinc finger protein 335 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |