Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACACAGGTGGTTCATGGGGCTGA[A/G]GCAGCGCAGTTGGTCCTCTGGGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602905 MIM: 605161 | ||||||||||||||||||||
Literature Links: |
KCNS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCNS1 - potassium voltage-gated channel modifier subfamily S member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WFDC5 - WAP four-disulfide core domain 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145652.3 | 354 | Missense Mutation | CTC,TTC | L,F 89 | NP_663627.1 | |
XM_011528601.1 | 354 | Missense Mutation | CTC,TTC | L,F 89 | XP_011526903.1 | |
XM_011528602.1 | 354 | Missense Mutation | CTC,TTC | L,F 89 | XP_011526904.1 |