Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATCATGAAAGTCGCCAGTGGCAG[C/G]ACCGCCACCGCCGCCGCGGGCCCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600349 | ||||||||||||||||||||
Literature Links: |
ID1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ID1 - inhibitor of DNA binding 1, HLH protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002165.3 | 126 | Missense Mutation | AGC,AGG | S,R 7 | NP_002156.2 | |
NM_181353.2 | 126 | Missense Mutation | AGC,AGG | S,R 7 | NP_851998.1 |
MIR3193 - microRNA 3193 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |