Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATTCTGCCATTTTGCTAGCACTG[A/G]TATGGCTCTTGGGTCCACCTACAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602995 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM189-UBE2V1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM189-UBE2V1 - TMEM189-UBE2V1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199203.2 | 692 | Missense Mutation | CCA,TCA | P,S 329 | NP_954673.1 |
UBE2V1 - ubiquitin conjugating enzyme E2 V1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032288.2 | 692 | Missense Mutation | CCA,TCA | P,S 106 | NP_001027459.1 | |
NM_001257393.1 | 692 | Missense Mutation | CCA,TCA | P,S 129 | NP_001244322.1 | |
NM_001257394.1 | 692 | Missense Mutation | CCA,TCA | P,S 62 | NP_001244323.1 | |
NM_001257395.1 | 692 | Missense Mutation | CCA,TCA | P,S 103 | NP_001244324.1 | |
NM_001257396.1 | 692 | Missense Mutation | CCA,TCA | P,S 64 | NP_001244325.1 | |
NM_001257397.1 | 692 | Missense Mutation | CCA,TCA | P,S 64 | NP_001244326.1 | |
NM_001257398.1 | 692 | Missense Mutation | CCA,TCA | P,S 64 | NP_001244327.1 | |
NM_001257399.1 | 692 | Missense Mutation | CCA,TCA | P,S 64 | NP_001244328.1 | |
NM_001282575.2 | 692 | Missense Mutation | CCA,TCA | P,S 50 | NP_001269504.1 | |
NM_001282576.1 | 692 | Missense Mutation | CCA,TCA | P,S 64 | NP_001269505.1 | |
NM_001282577.2 | 692 | Missense Mutation | CCA,TCA | P,S 50 | NP_001269506.1 | |
NM_001282578.2 | 692 | Missense Mutation | CCA,TCA | P,S 50 | NP_001269507.1 | |
NM_001282579.1 | 692 | Missense Mutation | CCA,TCA | P,S 64 | NP_001269508.1 | |
NM_001282580.1 | 692 | Intron | NP_001269509.1 | |||
NM_021988.5 | 692 | Missense Mutation | CCA,TCA | P,S 129 | NP_068823.2 | |
NM_022442.5 | 692 | Missense Mutation | CCA,TCA | P,S 62 | NP_071887.1 | |
NM_199144.2 | 692 | Missense Mutation | CCA,TCA | P,S 129 | NP_954595.1 |