Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTCTGCGGAATCAGGGAAGAGT[A/G]AAAAGGGTCAGCCACAGAATTCCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602184 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDUFV3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NDUFV3 - NADH:ubiquinone oxidoreductase subunit V3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001503.1 | 190 | Missense Mutation | AAA,GAA | K,E 42 | NP_001001503.1 | |
NM_021075.3 | 190 | Missense Mutation | AAA,GAA | K,E 42 | NP_066553.3 | |
XM_011529586.2 | 190 | Missense Mutation | AAA,GAA | K,E 42 | XP_011527888.1 | |
XM_017028359.1 | 190 | Missense Mutation | AAA,GAA | K,E 42 | XP_016883848.1 |