Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTAATTTTTTCTTAGCCGCAGCAG[A/C]AGCATTTCGATGGGGCAGTTTTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605296 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRWD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRWD1 - bromodomain and WD repeat domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007246.2 | 7112 | Intron | NP_001007247.1 | |||
NM_018963.4 | 7112 | Missense Mutation | GCT,TCT | A,S 2273 | NP_061836.2 | |
NM_033656.3 | 7112 | UTR 3 | NP_387505.1 | |||
XM_011529611.1 | 7112 | Missense Mutation | GCT,TCT | A,S 2225 | XP_011527913.1 | |
XM_011529612.2 | 7112 | Missense Mutation | GCT,TCT | A,S 1993 | XP_011527914.1 | |
XM_011529613.1 | 7112 | Missense Mutation | GCT,TCT | A,S 1284 | XP_011527915.1 | |
XM_017028373.1 | 7112 | Missense Mutation | GCT,TCT | A,S 2186 | XP_016883862.1 | |
XM_017028374.1 | 7112 | Missense Mutation | GCT,TCT | A,S 1284 | XP_016883863.1 | |
XM_017028375.1 | 7112 | Intron | XP_016883864.1 |
PSMG1 - proteasome assembly chaperone 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |