Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGACCCGGGACCCCTGCCGAGCGC[C/T]ACGCCGACGGCTTGGCGCTCGCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605668 MIM: 613770 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BACE2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BACE2 - beta-site APP-cleaving enzyme 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012105.4 | 629 | Missense Mutation | CAC,TAC | H,Y 56 | NP_036237.2 | |
NM_138991.2 | 629 | Missense Mutation | CAC,TAC | H,Y 56 | NP_620476.1 | |
NM_138992.2 | 629 | Missense Mutation | CAC,TAC | H,Y 56 | NP_620477.1 | |
XM_017028314.1 | 629 | Intron | XP_016883803.1 |
MIR3197 - microRNA 3197 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLAC4 - placenta specific 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |