Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTTCGAGGAGCGCCACATCCCGG[G/T]CGCCGCTTTCTTCGACATCGACCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602496 MIM: 180370 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MPST PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MPST - mercaptopyruvate sulfurtransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013436.2 | 324 | Missense Mutation | GGC,GTC | G,V 56 | NP_001013454.1 | |
NM_001130517.2 | 324 | Missense Mutation | GGC,GTC | G,V 56 | NP_001123989.1 | |
NM_021126.5 | 324 | Missense Mutation | GGC,GTC | G,V 76 | NP_066949.2 | |
XM_005261610.3 | 324 | Missense Mutation | GGC,GTC | G,V 56 | XP_005261667.1 | |
XM_011530195.1 | 324 | Missense Mutation | GGC,GTC | G,V 76 | XP_011528497.1 | |
XM_011530196.1 | 324 | Missense Mutation | GGC,GTC | G,V 56 | XP_011528498.1 |
TST - thiosulfate sulfurtransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |