Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACTTTCTGCAGAGCGTTGCTGAT[A/G]GGGCCTGTCCGTGGTCAAGGGCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616386 MIM: 609862 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCTD17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCTD17 - potassium channel tetramerization domain containing 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMPRSS6 - transmembrane protease, serine 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289000.1 | 1952 | Silent Mutation | CCC,CCT | P,P 728 | NP_001275929.1 | |
NM_001289001.1 | 1952 | Silent Mutation | CCC,CCT | P,P 706 | NP_001275930.1 | |
NM_153609.3 | 1952 | Silent Mutation | CCC,CCT | P,P 715 | NP_705837.1 | |
XM_011529989.2 | 1952 | Silent Mutation | CCC,CCT | P,P 584 | XP_011528291.1 |