Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGCTCAGTTGTCTCCTGGAAGAC[A/G]GGCGTCTTCCGAAACCAGGTAGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607750 MIM: 609900 MIM: 608993 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOBEC3C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APOBEC3C - apolipoprotein B mRNA editing enzyme catalytic subunit 3C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014508.2 | 259 | Silent Mutation | ACA,ACG | T,T 52 | NP_055323.2 |
APOBEC3D - apolipoprotein B mRNA editing enzyme catalytic subunit 3D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
APOBEC3F - apolipoprotein B mRNA editing enzyme catalytic subunit 3F | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |