Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTGTTTTTGGGGACTGCCATGTC[A/G]CAGGAGAAAGAGCCATTGGCACTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605165 | ||||||||||||||||||||
Literature Links: |
PATZ1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PATZ1 - POZ/BTB and AT hook containing zinc finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014323.2 | 2287 | Silent Mutation | TGC,TGT | C,C 588 | NP_055138.2 | |
NM_032050.1 | 2287 | Silent Mutation | TGC,TGT | C,C 542 | NP_114439.1 | |
NM_032051.1 | 2287 | Intron | NP_114440.1 | |||
NM_032052.1 | 2287 | UTR 3 | NP_114441.1 |
PIK3IP1-AS1 - PIK3IP1 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |