Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCACTTGTTTTTTGCTCCTCCAA[A/G]TCCACCTAATGGTGATAAGAGTGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605968 MIM: 605972 | ||||||||||||||||||||
Literature Links: |
RFPL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RFPL1 - ret finger protein like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021026.2 | 4394 | Intron | NP_066306.2 | |||
XM_011530298.2 | 4394 | Missense Mutation | AAT,AGT | N,S 258 | XP_011528600.1 | |
XM_011530299.2 | 4394 | Intron | XP_011528601.1 | |||
XM_017028900.1 | 4394 | Missense Mutation | AAT,AGT | N,S 258 | XP_016884389.1 | |
XM_017028901.1 | 4394 | Missense Mutation | AAT,AGT | N,S 258 | XP_016884390.1 | |
XM_017028902.1 | 4394 | Missense Mutation | AAT,AGT | N,S 258 | XP_016884391.1 |
RFPL1S - RFPL1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |