Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTCGAGTTTCACCACGAGCGCGGC[C/T]TGGTGAAGGTCAACGACAAAGAGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 151410 MIM: 605663 | ||||||||||||||||||||
Literature Links: |
BCR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BCR - BCR, RhoGEF and GTPase activating protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004327.3 | 1158 | Silent Mutation | CTG,TTG | L,L 188 | NP_004318.3 | |
NM_021574.2 | 1158 | Silent Mutation | CTG,TTG | L,L 188 | NP_067585.2 |
RSPH14 - radial spoke head 14 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |