Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCAGGGGCCTAGCCCTGGAAGCC[C/G]AGCTCCTTGTAGTTGGAGGCCATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 160000 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MB - myoglobin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005368.2 | 622 | Silent Mutation | CTC,CTG | L,L 150 | NP_005359.1 | |
NM_203377.1 | 622 | Silent Mutation | CTC,CTG | L,L 150 | NP_976311.1 | |
NM_203378.1 | 622 | Silent Mutation | CTC,CTG | L,L 150 | NP_976312.1 | |
XM_005261605.2 | 622 | Silent Mutation | CTC,CTG | L,L 150 | XP_005261662.1 |