Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCATGTCACTCAGCAGTTCCAGGC[C/T]CTAGATGGGGCAGCCAAACCGGGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC100652901 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC100652901 - uncharacterized LOC100652901 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRP5L - LDL receptor related protein 5 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135772.1 | 3870 | Silent Mutation | NP_001129244.1 | |||
NM_182492.2 | 3870 | Intron | NP_872298.1 | |||
XM_006724372.3 | 3870 | Missense Mutation | XP_006724435.1 | |||
XM_017029083.1 | 3870 | Missense Mutation | XP_016884572.1 | |||
XM_017029084.1 | 3870 | Missense Mutation | XP_016884573.1 | |||
XM_017029085.1 | 3870 | Missense Mutation | XP_016884574.1 | |||
XM_017029086.1 | 3870 | Missense Mutation | XP_016884575.1 | |||
XM_017029087.1 | 3870 | Missense Mutation | XP_016884576.1 | |||
XM_017029088.1 | 3870 | Missense Mutation | XP_016884577.1 |