Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTTCCTTTCGTATATGGCATGC[A/G]TCGTCTGCTCCCACTTCACTGGGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603392 MIM: 614479 | ||||||||||||||||||||
Literature Links: |
BIK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BIK - BCL2 interacting killer | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MCAT - malonyl-CoA-acyl carrier protein transacylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014507.3 | 1038 | UTR 3 | NP_055322.1 | |||
NM_173467.4 | 1038 | Missense Mutation | ACG,ATG | T,M 330 | NP_775738.3 | |
XM_011530156.2 | 1038 | Missense Mutation | ACG,ATG | T,M 165 | XP_011528458.1 |