Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAACGGAGGCTTTGACCTGGTGC[A/G]TCCGGCCTCCTGGCAGGAGACAGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607755 | ||||||||||||||||||||
Literature Links: |
CHKB-AS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHKB-AS1 - CHKB antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAPK8IP2 - mitogen-activated protein kinase 8 interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012324.4 | 508 | Missense Mutation | CAT,CGT | H,R 162 | NP_036456.1 | |
XM_011530679.2 | 508 | Missense Mutation | CAT,CGT | H,R 162 | XP_011528981.1 | |
XM_011530680.2 | 508 | Missense Mutation | CAT,CGT | H,R 162 | XP_011528982.1 | |
XM_011530681.2 | 508 | Missense Mutation | CAT,CGT | H,R 162 | XP_011528983.1 |