Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTCTGAACAGAGACCAGAGGCTC[A/G]CCTGGGTGAGTACCACTGCCTCCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
RRP7BP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RRP7BP - ribosomal RNA processing 7 homolog B, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SERHL2 - serine hydrolase-like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284334.1 | 830 | Intron | NP_001271263.1 | |||
NM_014509.4 | 830 | Intron | NP_055324.2 | |||
XM_017028738.1 | 830 | Missense Mutation | ACC,GCC | T,A 228 | XP_016884227.1 | |
XM_017028739.1 | 830 | Missense Mutation | ACC,GCC | T,A 201 | XP_016884228.1 |