Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCAGCAGCTGATCCGGGGAGTCC[C/T]CTGCAGGCGCCCCCACGGGCTTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607209 MIM: 602577 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CARD10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CARD10 - caspase recruitment domain family member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014550.3 | 2441 | Missense Mutation | GAG,GGG | E,G 813 | NP_055365.2 |
MFNG - MFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |