Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGAGGGCACCATGCAAAACGCGC[A/T]GGGCCTTGTCCCTCTCTGCAGCGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609030 MIM: 601180 MIM: 611151 | ||||||||||||||||||||
Literature Links: |
DGCR8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DGCR8 - DGCR8 microprocessor complex subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6816 - microRNA 6816 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RANBP1 - RAN binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278639.1 | 573 | Missense Mutation | AGG,TGG | R,W 26 | NP_001265568.1 | |
NM_001278640.1 | 573 | Intron | NP_001265569.1 | |||
NM_001278641.1 | 573 | Intron | NP_001265570.1 | |||
NM_002882.3 | 573 | Intron | NP_002873.1 | |||
XM_011530289.2 | 573 | Intron | XP_011528591.1 | |||
XM_011530290.2 | 573 | Intron | XP_011528592.1 | |||
XM_011530291.2 | 573 | Intron | XP_011528593.1 | |||
XM_017028890.1 | 573 | Intron | XP_016884379.1 | |||
XM_017028891.1 | 573 | Intron | XP_016884380.1 | |||
XM_017028892.1 | 573 | Intron | XP_016884381.1 |
TRMT2A - tRNA methyltransferase 2 homolog A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257994.1 | 573 | Missense Mutation | CAG,CTG | Q,L 126 | NP_001244923.1 | |
NM_022727.5 | 573 | Missense Mutation | CAG,CTG | Q,L 126 | NP_073564.3 | |
NM_182984.4 | 573 | Missense Mutation | CAG,CTG | Q,L 126 | NP_892029.2 | |
XM_011530139.2 | 573 | Missense Mutation | CAG,CTG | Q,L 126 | XP_011528441.1 | |
XM_011530141.1 | 573 | Intron | XP_011528443.1 | |||
XM_011530142.2 | 573 | Intron | XP_011528444.1 | |||
XM_017028770.1 | 573 | Missense Mutation | CAG,CTG | Q,L 126 | XP_016884259.1 |