Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603488 MIM: 610147 MIM: 609023 MIM: 610364 | ||||||||||||||||||||
Literature Links: |
AAMP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AAMP - angio associated migratory cell protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPBAR1 - G protein-coupled bile acid receptor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6513 - microRNA 6513 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PNKD - paroxysmal nonkinesigenic dyskinesia | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077399.2 | 327 | Intron | NP_001070867.1 | |||
NM_015488.4 | 327 | Silent Mutation | GAA,GAG | E,E 61 | NP_056303.3 | |
NM_022572.4 | 327 | Intron | NP_072094.1 | |||
XM_017003771.1 | 327 | Silent Mutation | GAA,GAG | E,E 61 | XP_016859260.1 | |
XM_017003772.1 | 327 | Intron | XP_016859261.1 |
TMBIM1 - transmembrane BAX inhibitor motif containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |