Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTGGGTTGGTCAGAGGGAAGAG[C/T]CATGGCAATCACAAAATTCGTGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610061 MIM: 608733 | ||||||||||||||||||||
Literature Links: |
DNAH7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAH7 - dynein axonemal heavy chain 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018897.2 | 6498 | Missense Mutation | NP_061720.2 | |||
XM_011511487.2 | 6498 | Missense Mutation | XP_011509789.1 | |||
XM_011511488.2 | 6498 | Missense Mutation | XP_011509790.1 | |||
XM_011511489.2 | 6498 | Missense Mutation | XP_011509791.1 | |||
XM_011511490.2 | 6498 | Missense Mutation | XP_011509792.1 | |||
XM_011511491.2 | 6498 | Intron | XP_011509793.1 | |||
XM_011511492.2 | 6498 | Intron | XP_011509794.1 | |||
XM_011511493.2 | 6498 | Intron | XP_011509795.1 | |||
XM_011511494.2 | 6498 | Intron | XP_011509796.1 | |||
XM_011511495.2 | 6498 | Intron | XP_011509797.1 | |||
XM_011511497.2 | 6498 | Missense Mutation | XP_011509799.1 | |||
XM_017004504.1 | 6498 | Missense Mutation | XP_016859993.1 |
SLC39A10 - solute carrier family 39 member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127257.1 | 6498 | Intron | NP_001120729.1 | |||
NM_020342.2 | 6498 | Intron | NP_065075.1 | |||
XM_005246689.4 | 6498 | Intron | XP_005246746.2 | |||
XM_011511504.2 | 6498 | Intron | XP_011509806.1 | |||
XM_011511505.2 | 6498 | Intron | XP_011509807.1 | |||
XM_011511506.2 | 6498 | Intron | XP_011509808.1 | |||
XM_011511507.2 | 6498 | Intron | XP_011509809.1 | |||
XM_017004522.1 | 6498 | Intron | XP_016860011.1 | |||
XM_017004523.1 | 6498 | Intron | XP_016860012.1 |