Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCAAAGAAGGCAAGCTCTTTATT[C/G]TTATCAAGCTTTGCTCTGTGGGCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 147700 | ||||||||||||||||||||
Literature Links: |
IDH1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IDH1 - isocitrate dehydrogenase (NADP(+)) 1, cytosolic | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282386.1 | 1407 | Intron | NP_001269315.1 | |||
NM_001282387.1 | 1407 | Missense Mutation | AAC,AAG | N,K 348 | NP_001269316.1 | |
NM_005896.3 | 1407 | Intron | NP_005887.2 |