Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTCTCCTTGCAGCAGCCCCTGTC[A/G]AAGGGAACAGGAAGCAGTCCACGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611583 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARID5A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARID5A - AT-rich interaction domain 5A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319085.1 | 124 | Missense Mutation | AAA,GAA | K,E 6 | NP_001306014.1 | |
NM_001319087.1 | 124 | Missense Mutation | AAA,GAA | K,E 6 | NP_001306016.1 | |
NM_001319092.1 | 124 | UTR 5 | NP_001306021.1 | |||
NM_001319093.1 | 124 | UTR 5 | NP_001306022.1 | |||
NM_001319094.1 | 124 | UTR 5 | NP_001306023.1 | |||
NM_001319096.1 | 124 | UTR 5 | NP_001306025.1 | |||
NM_212481.2 | 124 | Missense Mutation | AAA,GAA | K,E 6 | NP_997646.1 | |
XM_011510499.2 | 124 | Missense Mutation | AAA,GAA | K,E 16 | XP_011508801.1 | |
XM_017003183.1 | 124 | Missense Mutation | AAA,GAA | K,E 16 | XP_016858672.1 | |
XM_017003184.1 | 124 | UTR 5 | XP_016858673.1 | |||
XM_017003185.1 | 124 | UTR 5 | XP_016858674.1 |