Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCACCCCTCCCCACCCTCTGTCCC[A/G]CAGTGAGGACGGGACTCTACTGCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605096 MIM: 602877 | ||||||||||||||||||||
Literature Links: |
ANO7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANO7 - anoctamin 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001666.3 | 125 | Missense Mutation | ACA,GCA | T,A 22 | NP_001001666.1 | |
NM_001001891.3 | 125 | Missense Mutation | ACA,GCA | T,A 22 | NP_001001891.2 | |
XM_011511263.2 | 125 | Missense Mutation | ACA,GCA | T,A 22 | XP_011509565.1 | |
XM_011511267.2 | 125 | Missense Mutation | ACA,GCA | T,A 22 | XP_011509569.1 | |
XM_017004229.1 | 125 | Missense Mutation | ACA,GCA | T,A 22 | XP_016859718.1 | |
XM_017004230.1 | 125 | Intron | XP_016859719.1 |
PPP1R7 - protein phosphatase 1 regulatory subunit 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |