Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTCAACTTATTCCCTTCAATTC[A/G]AGTAACAGGTAAACAATGTTAATGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 186760 | ||||||||||||||||||||
Literature Links: |
CD28 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD28 - CD28 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243077.1 | 266 | Missense Mutation | CAA,CGA | Q,R 15 | NP_001230006.1 | |
NM_001243078.1 | 266 | Missense Mutation | CAA,CGA | Q,R 15 | NP_001230007.1 | |
NM_006139.3 | 266 | Missense Mutation | CAA,CGA | Q,R 15 | NP_006130.1 | |
XM_011512194.2 | 266 | Intron | XP_011510496.1 | |||
XM_011512195.2 | 266 | Intron | XP_011510497.1 | |||
XM_011512197.2 | 266 | Missense Mutation | CAA,CGA | Q,R 15 | XP_011510499.1 |