Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCACAGGGTTAGACGTATCTCGCC[A/G]AAAGGCTGGGCTTGTCTCCCGATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 186910 | ||||||||||||||||||||
Literature Links: |
CD8A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD8A - CD8a molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145873.1 | 1723 | Missense Mutation | TCG,TTG | S,L 231 | NP_001139345.1 | |
NM_001768.6 | 1723 | Missense Mutation | TCG,TTG | S,L 231 | NP_001759.3 | |
NM_171827.3 | 1723 | Missense Mutation | TCG,TTG | S,L 194 | NP_741969.1 |
RMND5A - required for meiotic nuclear division 5 homolog A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |