Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCCAGTCCAAGGCTTCTTCTCT[C/T]CTTTGTTTTTGAAAGTTTTGCTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SRBD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SRBD1 - S1 RNA binding domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018079.4 | 2511 | Missense Mutation | AAG,GAG | K,E 961 | NP_060549.4 | |
XM_011532946.1 | 2511 | Missense Mutation | AAG,GAG | K,E 945 | XP_011531248.1 | |
XM_017004417.1 | 2511 | Intron | XP_016859906.1 | |||
XM_017004418.1 | 2511 | Missense Mutation | AAG,GAG | K,E 464 | XP_016859907.1 |