Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGCCTGCTCCTCTTCCTCTGTGG[A/G]GTAGTGGCGCAGGAGGTTGGCTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613403 | ||||||||||||||||||||
Literature Links: |
TMEM127 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM127 - transmembrane protein 127 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193304.2 | 500 | Missense Mutation | NP_001180233.1 | |||
NM_017849.3 | 500 | Missense Mutation | NP_060319.1 | |||
XM_017004450.1 | 500 | UTR 5 | XP_016859939.1 | |||
XM_017004451.1 | 500 | Missense Mutation | XP_016859940.1 | |||
XM_017004452.1 | 500 | Missense Mutation | XP_016859941.1 |