Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAGAGGAAGAATATATTTTGACA[A/G]TTATCGGCGCTGTGTCAGCAGGTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612515 MIM: 609525 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCAF17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCAF17 - DDB1 and CUL4 associated factor 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164821.1 | 188 | Missense Mutation | AAT,AGT | N,S 70 | NP_001158293.1 | |
NM_025000.3 | 188 | Missense Mutation | AAT,AGT | N,S 70 | NP_079276.2 | |
XM_006712767.1 | 188 | UTR 5 | XP_006712830.1 | |||
XM_006712768.1 | 188 | UTR 5 | XP_006712831.1 | |||
XM_011511881.1 | 188 | Missense Mutation | AAT,AGT | N,S 70 | XP_011510183.1 | |
XM_011511882.1 | 188 | Missense Mutation | AAT,AGT | N,S 70 | XP_011510184.1 | |
XM_017004995.1 | 188 | Missense Mutation | AAT,AGT | N,S 70 | XP_016860484.1 | |
XM_017004996.1 | 188 | Missense Mutation | AAT,AGT | N,S 70 | XP_016860485.1 | |
XM_017004997.1 | 188 | Missense Mutation | AAT,AGT | N,S 70 | XP_016860486.1 | |
XM_017004998.1 | 188 | UTR 5 | XP_016860487.1 | |||
XM_017004999.1 | 188 | Missense Mutation | AAT,AGT | N,S 70 | XP_016860488.1 | |
XM_017005000.1 | 188 | Missense Mutation | AAT,AGT | N,S 70 | XP_016860489.1 | |
XM_017005001.1 | 188 | Missense Mutation | AAT,AGT | N,S 70 | XP_016860490.1 | |
XM_017005002.1 | 188 | UTR 5 | XP_016860491.1 |
METTL8 - methyltransferase like 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321154.1 | 188 | Intron | NP_001308083.1 | |||
NM_001321155.1 | 188 | Intron | NP_001308084.1 | |||
NM_001321156.1 | 188 | Intron | NP_001308085.1 | |||
NM_001321157.1 | 188 | Intron | NP_001308086.1 | |||
NM_001321158.1 | 188 | Intron | NP_001308087.1 | |||
NM_001321159.1 | 188 | Intron | NP_001308088.1 | |||
NM_001321160.1 | 188 | Intron | NP_001308089.1 | |||
NM_001321161.1 | 188 | Intron | NP_001308090.1 | |||
NM_001321162.1 | 188 | Intron | NP_001308091.1 | |||
NM_024770.4 | 188 | Intron | NP_079046.2 | |||
XM_011511875.2 | 188 | Intron | XP_011510177.1 | |||
XM_017004984.1 | 188 | Intron | XP_016860473.1 | |||
XM_017004985.1 | 188 | Intron | XP_016860474.1 | |||
XM_017004986.1 | 188 | Intron | XP_016860475.1 | |||
XM_017004987.1 | 188 | Intron | XP_016860476.1 | |||
XM_017004988.1 | 188 | Intron | XP_016860477.1 | |||
XM_017004989.1 | 188 | Intron | XP_016860478.1 |