Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAATATTTAGCAGCTAAGTCCTGA[A/G]AACATTCATCATTCCCTTCACTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAD51AP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAD51AP2 - RAD51 associated protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099218.2 | 2852 | Missense Mutation | TCT,TTT | S,F 943 | NP_001092688.1 | |
NM_001321233.1 | 2852 | Missense Mutation | TCT,TTT | S,F 934 | NP_001308162.1 | |
XM_011533084.1 | 2852 | Missense Mutation | TCT,TTT | S,F 943 | XP_011531386.1 |