Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTATTTTTTGCTTTCCTTGCTG[A/T]CTTTCCCTTTGTCTTCCTTTTCTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603415 | ||||||||||||||||||||
Literature Links: |
LOC101929680 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101929680 - uncharacterized LOC101929680 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCN9A - sodium voltage-gated channel alpha subunit 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002977.3 | 6220 | Missense Mutation | GAC,GTC | D,V 1971 | NP_002968.1 | |
XM_005246757.2 | 6220 | Missense Mutation | GAC,GTC | D,V 1982 | XP_005246814.1 | |
XM_011511616.2 | 6220 | Missense Mutation | GAC,GTC | D,V 1982 | XP_011509918.1 | |
XM_011511617.2 | 6220 | Missense Mutation | GAC,GTC | D,V 1982 | XP_011509919.1 | |
XM_011511618.2 | 6220 | Missense Mutation | GAC,GTC | D,V 1971 | XP_011509920.1 | |
XM_011511619.2 | 6220 | Intron | XP_011509921.1 | |||
XM_017004668.1 | 6220 | Missense Mutation | GAC,GTC | D,V 1853 | XP_016860157.1 | |
XM_017004669.1 | 6220 | Missense Mutation | GAC,GTC | D,V 1734 | XP_016860158.1 |