Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTCAGGCATAGAATTCCTCCTGTT[C/T]GGTGGGCTTCTGGTAGGCCCCGCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186355 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SDC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SDC1 - syndecan 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006946.1 | 1301 | Missense Mutation | AAA,GAA | K,E 304 | NP_001006947.1 | |
NM_002997.4 | 1301 | Missense Mutation | AAA,GAA | K,E 304 | NP_002988.3 | |
XM_005262620.4 | 1301 | Missense Mutation | AAA,GAA | K,E 328 | XP_005262677.1 | |
XM_005262621.3 | 1301 | Missense Mutation | AAA,GAA | K,E 299 | XP_005262678.3 | |
XM_005262622.2 | 1301 | Missense Mutation | AAA,GAA | K,E 286 | XP_005262679.1 |