Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCAGCTGCCCTCCAGCACATTGA[C/G]GGAGTGAATTTCAGTGAGGTGGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123670 MIM: 123680 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRYGB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRYGB - crystallin gamma B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005210.3 | 406 | Missense Mutation | CTC,GTC | L,V 125 | NP_005201.2 | |
XM_017003402.1 | 406 | Missense Mutation | CTC,GTC | L,V 127 | XP_016858891.1 |
CRYGC - crystallin gamma C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100507443 - uncharacterized LOC100507443 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |