Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGGGCAATCTTCGCCACAAAAGC[A/G]CCTAAATAAGAAATTATTGATTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612003 MIM: 603208 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GIGYF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GIGYF2 - GRB10 interacting GYF protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001103146.1 | 363 | Intron | NP_001096616.1 | |||
NM_001103147.1 | 363 | Intron | NP_001096617.1 | |||
NM_001103148.1 | 363 | Intron | NP_001096618.1 | |||
NM_015575.3 | 363 | Intron | NP_056390.2 |
KCNJ13 - potassium voltage-gated channel subfamily J member 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172416.1 | 363 | Missense Mutation | CGC,TGC | R,C 76 | NP_001165887.1 | |
NM_001172417.1 | 363 | Silent Mutation | GGC,GGT | G,G 74 | NP_001165888.1 | |
NM_002242.4 | 363 | Silent Mutation | GGC,GGT | G,G 154 | NP_002233.2 |