Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGAGCAGTATGGCCAGTGTGTGG[C/T]GGCCAAGCCGGAATCCTGGCAGCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616978 MIM: 602000 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHCHD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHCHD5 - coiled-coil-helix-coiled-coil-helix domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304353.1 | 1113 | UTR 5 | NP_001291282.1 | |||
NM_001304354.1 | 1113 | UTR 5 | NP_001291283.1 | |||
NM_032309.3 | 1113 | Missense Mutation | GCG,GTG | A,V 24 | NP_115685.1 | |
XM_005263824.3 | 1113 | Missense Mutation | GCG,GTG | A,V 24 | XP_005263881.1 |
POLR1B - RNA polymerase I subunit B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |