Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTTTGGGTGATATCTAAGCAGAC[A/G]TTTCCCTCTGAAGTGGTTGGCAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608952 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RIF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RIF1 - replication timing regulatory factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177663.1 | 422 | Silent Mutation | ACA,ACG | T,T 127 | NP_001171134.1 | |
NM_001177664.1 | 422 | Silent Mutation | ACA,ACG | T,T 127 | NP_001171135.1 | |
NM_001177665.1 | 422 | Silent Mutation | ACA,ACG | T,T 127 | NP_001171136.1 | |
NM_018151.4 | 422 | Silent Mutation | ACA,ACG | T,T 127 | NP_060621.3 | |
XM_005246665.3 | 422 | Silent Mutation | ACA,ACG | T,T 127 | XP_005246722.1 | |
XM_011511395.2 | 422 | Silent Mutation | ACA,ACG | T,T 127 | XP_011509697.1 | |
XM_017004422.1 | 422 | Silent Mutation | ACA,ACG | T,T 127 | XP_016859911.1 | |
XM_017004423.1 | 422 | Silent Mutation | ACA,ACG | T,T 127 | XP_016859912.1 | |
XM_017004424.1 | 422 | Silent Mutation | ACA,ACG | T,T 91 | XP_016859913.1 | |
XM_017004425.1 | 422 | Silent Mutation | ACA,ACG | T,T 127 | XP_016859914.1 |