Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGGCCAGTGTGACTCACCGATG[A/G]CCAGGTGTGCAGAGGCAACATGGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602878 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC30A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC30A3 - solute carrier family 30 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318949.1 | 1056 | Missense Mutation | GCC,GTC | A,V 333 | NP_001305878.1 | |
NM_001318950.1 | 1056 | Missense Mutation | GCC,GTC | A,V 325 | NP_001305879.1 | |
NM_001318951.1 | 1056 | Missense Mutation | GCC,GTC | A,V 325 | NP_001305880.1 | |
NM_003459.4 | 1056 | Missense Mutation | GCC,GTC | A,V 338 | NP_003450.2 | |
XM_005264547.1 | 1056 | Missense Mutation | CCA,TCA | P,S 287 | XP_005264604.1 | |
XM_005264548.3 | 1056 | Missense Mutation | CCA,TCA | P,S 282 | XP_005264605.1 | |
XM_006712100.2 | 1056 | Missense Mutation | CCA,TCA | P,S 238 | XP_006712163.1 | |
XM_011533102.2 | 1056 | Missense Mutation | GCC,GTC | A,V 265 | XP_011531404.1 | |
XM_011533103.2 | 1056 | Missense Mutation | GCC,GTC | A,V 224 | XP_011531405.1 | |
XM_017004873.1 | 1056 | Missense Mutation | CCA,TCA | P,S 274 | XP_016860362.1 | |
XM_017004874.1 | 1056 | Missense Mutation | CCA,TCA | P,S 274 | XP_016860363.1 | |
XM_017004875.1 | 1056 | Missense Mutation | CCA,TCA | P,S 265 | XP_016860364.1 |